Learn More
Perkin Elmer US LLC 81-Position Sample Tray for Series 200 Autosampler
Catalog No. 502111759
Encompass_Preferred
PerkinElmer has researched, engineered, and developed the Series 200 Autosampler to ensure precision, flexibility, reliability, and ease-of-use. Our unique XYZ design allows you to take advantage of the built-in extra capabilities including dilution, derivatization, and standard addition all without additional hardware or cost.
Catalog No. 50-211-1759 Supplier Perkin Elmer US LLC Supplier No. N2930044
PerkinElmer has researched, engineered, and developed the Series 200 Autosampler to ensure precision, flexibility, reliability, and ease-of-use. Our unique XYZ design allows you to take advantage of the built-in extra capabilities including dilution, derivatization, and standard addition all without additional hardware or cost.