Learn More
IBL America Amyloid Beta (N) (82E1) Aβ Anti-Human Mouse IgG MoAb
Supplier:  IBL America 10323
Encompass_Preferred
For research use only, not for use in diagnostic procedures.This antibody can be stained in formalin fixed paraffin embedded tissues after formicacid treatment*1 by several Immunohistochemical techniques such as Avidin BitionComplex (ABC) Method. The optimal dilution is about 1g /ml, however, the dilutionrate should be optimized by each laboratories.*1: rinsing by running water after formic acid treatment for 5 minutes followingde-paraffin. This antibody can be used for western blotting in concentration of about 1g /mL. This antibody can be used for Immunoprecipitation.
Catalog No. NC9747589