Learn More
IBL America Amyloid Beta (N) (82E1) Aβ Anti-Human Mouse IgG MoAb Biotin
Supplier:  IBL America 10326
Encompass_Preferred
For research use only, not for use in diagnostic procedures.This antibody can be stained in formalin fixed paraffin embedded tissues after formicacid treatment*1 by several Immunohistochemical techniques such as Avidin BiotinComplex (ABC) Method. The optimal dilution is 1g /mL, however, the dilution rateshould be optimized by each laboratories. *1: rinsing by running water after formic acid treatment for 5 minutes followingde-paraffin. This antibody can be used for western blotting in concentration of 1g /mL.
Catalog No. NC9747586