Learn More
eMolecules 83905-01-5 | Azithromycin | Medchem Express | MFCD03266623 | 748.996 | C38H72N2O12 | 98.000 | CC[C@H]1OC(=O)[C@H](C)[C@@H](O[C@H]2C[C@@](C)(OC)[C@@H](O)[C@H](C)O2)[C@H](C)[C@@H](O[C@@H]2O[C@H](C)C[C@@H]([C@H]2O)N(C)C)[C@](C)(O)C[C@@H](C)CN(C)[C@H](C)[C@@H](O)[C@]1(C)O | 100mg | 446268549
Shop All eMolecules Inc. Products