Learn More
Cole-Parmer Oakton Conductivity and TDS Standards Pack, 84 S, 1413 S, 12.88 mS, and 111.8 mS; 60-mL of each
Catalog No. NC2152121 Shop All Cole-Parmer Products
Encompass_Preferred
For your convenience, calibration standards, sensor heads, and sensor cartridges are available for your Oakton meters.
This item is not returnable. View return policy
Catalog No. NC2152121 Supplier Cole-Parmer Supplier No. 3566109
For your convenience, calibration standards, sensor heads, and sensor cartridges are available for your Oakton meters.