Learn More
eMolecules​ 84-58-2 | 2,3-Dichloro-5,6-dicyano-1,4-benzoquinone | Chem-Impex | MFCD00001593 | 227.000 | C8Cl2N2O2 | 98.000 | ClC1=C(Cl)C(=O)C(C#N)=C(C#N)C1=O | 250g | 112756269
Catalog No. 5000567116 Shop All eMolecules Inc. Products
Encompass_Preferred
2,3-Dichloro-5,6-dicyano-1,4-benzoquinone | Chem-Impex | 84-58-2 | MFCD00001593 | 227.000 | C8Cl2N2O2 | 98.000 | ClC1=C(Cl)C(=O)C(C#N)=C(C#N)C1=O | 250g | 112756269
Catalog No. 50-005-67116 Supplier eMolecules​ Supplier No. 112756269
2,3-Dichloro-5,6-dicyano-1,4-benzoquinone | Chem-Impex | 84-58-2 | MFCD00001593 | 227.000 | C8Cl2N2O2 | 98.000 | ClC1=C(Cl)C(=O)C(C#N)=C(C#N)C1=O | 250g | 112756269