Learn More
Medchemexpress LLC 84-B10 | 698346-43-9 | 99.6% | 473.44 | 1 ML
Supplier: Medchemexpress LLC HY4430710MM/1ML
84-B10 is a 3-phenylglutaric acid derivative. It inhibits cisplatin-induced tubular ferroptosis, attenuates mitochondrial damage and oxidative stress, and ameliorates acute kidney injury (AKI).
- Inhibits cisplatin-induced tubular epithelial cell ferroptosis.
- Restores cisplatin-induced mitochondrial structural damage and dysfunction.
- Attenuates mtROS-induced oxidative stress in cisplatin-induced AKI.
- Ameliorates cisplatin-induced acute kidney injury in mice.