Learn More
HANNA INSTRUMENTS INC 84 uS/cm Conductivity Standard (500mL Bottle)
SDP
Catalog No. NC1812380
Encompass
HI7033L is a premium quality 84 µS/cm conductivity standard that is prepared against a NIST traceable potassium chloride solution. Hanna conductivity solutions have the lot number and expiration date clearly marked on the label and are air tight with a tamper-proof seal to ensure the quality of the solution. Hanna's line of conductivity standards have been specially formulated to have an expiration of 5 years from the date of manufacture for an unopened bottle.Temperature chart of the actual µS/cm value at various temperatures printed on each bottle.Air tight bottle with tamper-proof seal to ensure quality.
Catalog No. NC1812380 Supplier HANNA INSTRUMENTS INC Supplier No. HI7033L
HI7033L is a premium quality 84 µS/cm conductivity standard that is prepared against a NIST traceable potassium chloride solution. Hanna conductivity solutions have the lot number and expiration date clearly marked on the label and are air tight with a tamper-proof seal to ensure the quality of the solution. Hanna's line of conductivity standards have been specially formulated to have an expiration of 5 years from the date of manufacture for an unopened bottle.Temperature chart of the actual µS/cm value at various temperatures printed on each bottle.Air tight bottle with tamper-proof seal to ensure quality.