Learn More
Enterprise Technology Solutions Laboratory Apron 35 X 45 Black One Size Fits Most Rubber
SDP
Catalog No. 502040821
Encompass_Preferred
Strong, impermeable apron is great for use in laboratories. It's a sturdy choice to protect the clothing of workers from chemical splashes. Stitched completely on all sides, with four aluminum grommets and three pre-tied 30" nylon tie strings. Removable, reusable ties provide versatility. Rubber coated material provides protective strength.
Catalog No. 50-204-0821 Supplier Enterprise Technology Solutions Supplier No. NSN6345023
Strong, impermeable apron is great for use in laboratories. It's a sturdy choice to protect the clothing of workers from chemical splashes. Stitched completely on all sides, with four aluminum grommets and three pre-tied 30" nylon tie strings. Removable, reusable ties provide versatility. Rubber coated material provides protective strength.