Learn More
Enterprise Technology Solutions Safety Helmet, White
SDP
Catalog No. 502041165
Encompass_Preferred
ABILITYONE 8415009353139 Skilcraft Cap style safety helmet is molded from high density polyethylene. Features short peak and trim profile. 4-point woven nylon suspension. Easy Quick-Slide sizing adjusts for head sizes 6 1/2" - 8".
Catalog No. 50-204-1165 Supplier Enterprise Technology Solutions Supplier No. NSN9353139
ABILITYONE 8415009353139 Skilcraft Cap style safety helmet is molded from high density polyethylene. Features short peak and trim profile. 4-point woven nylon suspension. Easy Quick-Slide sizing adjusts for head sizes 6 1/2" - 8".