Learn More
Aviva Systems Biology Corp HMCES Antibody - N-terminal region (ARP57366_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP57366_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against 8430410A17Rik. It was validated on Western Blot. . Alternate names: C85376ApplicationWB. Species reactivity: Cow, Dog, Horse, Human, Mouse, Pig, Rabbit, Rat
Catalog No. 50-286-302