Learn More
eMolecules​ 84434-11-7 | TPO-L | ChemScene | MFCD08692488 | 316.337 | C18H21O3P | 97.000 | CCOP(=O)(C(=O)c1c(C)cc(C)cc1C)c1ccccc1 | 1000g | 801483984
Catalog No. 5000482203 Shop All eMolecules Inc. Products
Encompass_Preferred
TPO-L | ChemScene | 84434-11-7 | MFCD08692488 | 316.337 | C18H21O3P | 97.000 | CCOP(=O)(C(=O)c1c(C)cc(C)cc1C)c1ccccc1 | 1000g | 801483984
Catalog No. 50-004-82203 Supplier eMolecules​ Supplier No. 801483984
TPO-L | ChemScene | 84434-11-7 | MFCD08692488 | 316.337 | C18H21O3P | 97.000 | CCOP(=O)(C(=O)c1c(C)cc(C)cc1C)c1ccccc1 | 1000g | 801483984