Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Lck Inhibitor 10mM | 847950-09-8 | 1 ML
SDP

Catalog No. 5000230398
Encompass_Preferred

Lck inhibitor is a small-molecule kinase inhibitor (CAS 847950-09-8) that potently inhibits Lck and shows activity against Lyn, Src, and Syk. It is supplied for research use and is soluble in DMSO for stock preparation; storage recommendations include -20°C for powder and -80°C for solutions.

  • Potent Lck inhibition (IC50 7 nM).
  • Activity against Lyn, Src, and Syk.
  • Molecular weight 530.62 g/mol; formula C31H30N8O.
  • High DMSO solubility for stock preparation.
  • Storage: powder at -20°C; solutions at -80°C.

Catalog No. 50-002-30398 Supplier Medchemexpress LLC Supplier No. HY1207210MM 1ML
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Lck inhibitor is a small-molecule kinase inhibitor (CAS 847950-09-8) that potently inhibits Lck and shows activity against Lyn, Src, and Syk. It is supplied for research use and is soluble in DMSO for stock preparation; storage recommendations include -20°C for powder and -80°C for solutions.

  • Potent Lck inhibition (IC50 7 nM).
  • Activity against Lyn, Src, and Syk.
  • Molecular weight 530.62 g/mol; formula C31H30N8O.
  • High DMSO solubility for stock preparation.
  • Storage: powder at -20°C; solutions at -80°C.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.