Learn More
Fisherbrand™ 850 Homogenizer Accessory, Motor Stand Assembly
GREENER_CHOICE
Catalog No. 15340172 Shop All Fisherbrand Products
Change view
Click to view available options
For Use With (Equipment):
Fisher Scientific 850 Homogenizer
Catalog No. For Use With (Equipment)
15-340-172 Fisher Scientific 850 Homogenizer
1 options
Catalog No. 15-340-172
Add to Cart
Add to Cart
Fisher Scientific™ 850 Homogenizer Accessory, Motor Stand Assembly designed to retain the Fisher Scientific 850 Homogenizer for post mounted operation.
  • Includes a baseplate and post; does not include motor clamp
  • Base dimensions: 12.7 x 20.3cm (5 x 8 in.) stainless steel base
  • Post dimensions: 48.2cm (19 in.) tall by 9.52mm (0.375 in.) diameter stainless steel support post
Specifications
Product Type Motor Stand Assembly
Material Stainless Steel
For Use With (Equipment) Fisher Scientific 850 Homogenizer
Includes Baseplate and post