Learn More
Bellco Glass, Inc. 850cm² Roller Bottle
SDP
Catalog No. 502330542
Encompass_Preferred
DescriptionLarge knurled cap for easy gripVented cap for open systemsVent caps feature 0.22µm hydrophobic membraneManufactured from medical grade virgin polystyreneTissue Culture Treated Roller Bottles have uniform hydrophilic surface ideal for consistent cell attachmentEasy-to-read black printed graduationsPackaged sterile in individual bags Pack Description1/BagCase Quantity12SterileYesNon-pyrogenicYesPrimary ColorBottle: ClearSecondary ColorCap: GreenPrimary MaterialBottle: PolystyreneSecondary MaterialCap: HDPEMaximum Volume2000mLGraduatedYesGraduation Increments (Subdivisions)50mLTissue Culture TreatedYesCell Growth Area850cm²RNase/DNase FreeYes
Catalog No. 50-233-0542 Supplier Bellco Glass, Inc. Supplier No. 229385000
DescriptionLarge knurled cap for easy gripVented cap for open systemsVent caps feature 0.22µm hydrophobic membraneManufactured from medical grade virgin polystyreneTissue Culture Treated Roller Bottles have uniform hydrophilic surface ideal for consistent cell attachmentEasy-to-read black printed graduationsPackaged sterile in individual bags Pack Description1/BagCase Quantity12SterileYesNon-pyrogenicYesPrimary ColorBottle: ClearSecondary ColorCap: GreenPrimary MaterialBottle: PolystyreneSecondary MaterialCap: HDPEMaximum Volume2000mLGraduatedYesGraduation Increments (Subdivisions)50mLTissue Culture TreatedYesCell Growth Area850cm²RNase/DNase FreeYes