Learn More
CELLTREAT Scientific Products 850cm² Roller Bottle, PET, Molded Graduations, Non-Vented Cap, Sterile, Case Quantity 18
SDP
Catalog No. 5000670027
Encompass_Preferred
PET Roller Bottles are naturally hydrophilic, suitable for most adherent cell culture
PET material is impact resistant
Ink-free, molded graduations
Catalog No. 50-006-70027 Supplier CELLTREAT Scientific Products Supplier No. 229334
PET Roller Bottles are naturally hydrophilic, suitable for most adherent cell culture
PET material is impact resistant
Ink-free, molded graduations