Learn More
AVANTOR MASTERFLEX (VWR LLC) Masterflex 230 VAC power cord with NEMA 6-15P plug, 6-ft long
Catalog No. NC1243249
Encompass
Choose an interchangeable cord/plug assembly for 115 or 230 VAC operation. Each 6-ft (1.8 m) cord and moded plug assembly features an IEC 320 appliance connector and rated for 10 A at 250 VAC.
Catalog No. NC1243249 Supplier AVANTOR MASTERFLEX (VWR LLC) Supplier No. MFLX5000178
Choose an interchangeable cord/plug assembly for 115 or 230 VAC operation. Each 6-ft (1.8 m) cord and moded plug assembly features an IEC 320 appliance connector and rated for 10 A at 250 VAC.