Learn More
Ace Glass, Inc. Complete 60mm OD vacuum trap, #15 threaded side arm and innertube
SDP
Catalog No. NC0759791
Encompass_Preferred
60mm O.D. vacuum trap consists of body with Ace-Thred, adjustable inner tube, and nylon bushing with FETFE o-ring. Has #15 threaded side arm. For use with PTFE Ace-Safe connectors, to allow safe, easy tubing connect-disconnect.
Catalog No. NC0759791 Supplier Ace Glass, Inc. Supplier No. 874647
60mm O.D. vacuum trap consists of body with Ace-Thred, adjustable inner tube, and nylon bushing with FETFE o-ring. Has #15 threaded side arm. For use with PTFE Ace-Safe connectors, to allow safe, easy tubing connect-disconnect.