Learn More
Perkin Elmer US LLC 8+8 Cell Changer Kit for the Linear Cell Changer
Catalog No. 502104137
Encompass_Preferred
PerkinElmer offers three cell changer kits with the Automatic Linear Transport (B2000186). Each kit includes an auxiliary reference position and additionall cell holders. These kits are: 5+5 Long Path Cell Changer Kit (N1010569) 8+8 Automatic Water-Thermostattable Cell Changer kit. 8+1 Automatic Wather-Thermostattable Cell Changer kit. These kits are compatible with the LAMBDA 20/40/20Bio/40Bio/40P/25/35/45 spectrometers.
Catalog No. 50-210-4137 Supplier Perkin Elmer US LLC Supplier No. N1010568
Add to Cart
Add to Cart
Request a Quote
PerkinElmer offers three cell changer kits with the Automatic Linear Transport (B2000186). Each kit includes an auxiliary reference position and additionall cell holders. These kits are: 5+5 Long Path Cell Changer Kit (N1010569) 8+8 Automatic Water-Thermostattable Cell Changer kit. 8+1 Automatic Wather-Thermostattable Cell Changer kit. These kits are compatible with the LAMBDA 20/40/20Bio/40Bio/40P/25/35/45 spectrometers.