Learn More
eMolecules​ 887144-94-7 | 1-(trifluoromethyl)-3H-1??,2-benziodaoxol-3-one | Pharmablock | MFCD18800706 | 316.018 | C8H4F3IO2 | 97.000 | FC(F)(F)[I]1OC(=O)c2ccccc12 | 25g | 551095437
Catalog No. 5000490277 Shop All eMolecules Inc. Products
Encompass_Preferred
1-(trifluoromethyl)-3H-1??,2-benziodaoxol-3-one | Pharmablock | 887144-94-7 | MFCD18800706 | 316.018 | C8H4F3IO2 | 97.000 | FC(F)(F)[I]1OC(=O)c2ccccc12 | 25g | 551095437
Catalog No. 50-004-90277 Supplier eMolecules​ Supplier No. 551095437
1-(trifluoromethyl)-3H-1??,2-benziodaoxol-3-one | Pharmablock | 887144-94-7 | MFCD18800706 | 316.018 | C8H4F3IO2 | 97.000 | FC(F)(F)[I]1OC(=O)c2ccccc12 | 25g | 551095437