Learn More
Andwin Scientific 89094-680/50ML 200/CS VWR8909
SDP
Catalog No. NC9939722
Encompass_Preferred
The disposable pipetting reservoirs are made for multichannel pipetting applications. This product is compatible with any type of pipettors that are up to 12-channel width. This polystyrene reservoir has a unique trough construction, graduation marks on interior walls, and pour-off spouts on each corner. It has a volume capacity of 50 mL and has a dimension of 14.6 x 8.3 x 3 cm. This product is sold in a case of 200 white sterile reservoirs.
Catalog No. NC9939722 Supplier Andwin Scientific Supplier No. 89094680
The disposable pipetting reservoirs are made for multichannel pipetting applications. This product is compatible with any type of pipettors that are up to 12-channel width. This polystyrene reservoir has a unique trough construction, graduation marks on interior walls, and pour-off spouts on each corner. It has a volume capacity of 50 mL and has a dimension of 14.6 x 8.3 x 3 cm. This product is sold in a case of 200 white sterile reservoirs.