Learn More
MSC Scotch 890MSR Tape, 24 mm x 55 m, 1 Roll
Catalog No. 501299903
Encompass
Filament tape offers high tensile strength, reinforced with continuous fiberglass-yarn filaments. It features a black polyester (PET) backing for excellent abrasion, moisture, and scuff resistance. The modified synthetic rubber adhesive ensures strong initial adhesion, especially to oily metal surfaces, providing long-term holding power.
  • Reinforced with continuous fiberglass-yarn filaments for very high tensile strength
  • Polyester backing provides excellent abrasion, moisture, and scuff resistance
  • Modified synthetic rubber adhesive for strong adhesion to various surfaces, including oily metals
  • Meets ASTM D5330 Type IV for black filament tape
  • Excellent aging performance before and after application
  • Printable by the print seal process for customization
  • Suitable for bundling, strapping, and reinforcing applications
Catalog No. 50-129-9903 Supplier MSC Supplier No. 78270766
Filament tape offers high tensile strength, reinforced with continuous fiberglass-yarn filaments. It features a black polyester (PET) backing for excellent abrasion, moisture, and scuff resistance. The modified synthetic rubber adhesive ensures strong initial adhesion, especially to oily metal surfaces, providing long-term holding power.
  • Reinforced with continuous fiberglass-yarn filaments for very high tensile strength
  • Polyester backing provides excellent abrasion, moisture, and scuff resistance
  • Modified synthetic rubber adhesive for strong adhesion to various surfaces, including oily metals
  • Meets ASTM D5330 Type IV for black filament tape
  • Excellent aging performance before and after application
  • Printable by the print seal process for customization
  • Suitable for bundling, strapping, and reinforcing applications