Learn More
MSC SCOTCH FILAMENT TAPE
Catalog No. 502734838
Encompass
Filament & Strapping Tape; Type: Filament Tape ; Color: Clear ; Width (mm - 2 Decimals): 18.00 ; Thickness (mil): 6.0000 ; Tape Number: 893 ; Length (Meters): 55.00
Catalog No. 50-273-4838 Supplier MSC Supplier No. 53354130
Filament & Strapping Tape; Type: Filament Tape ; Color: Clear ; Width (mm - 2 Decimals): 18.00 ; Thickness (mil): 6.0000 ; Tape Number: 893 ; Length (Meters): 55.00