Learn More
MSC Mechanix Wear Chemical-Resistant Gloves, Sandy Nitrile-Coated, HPPE, Medium, 1/Pr.
Catalog No. 5000684868
Encompass
These chemical-resistant impact gloves are constructed from a high-dexterity 18-gauge HPPE and glass fiber blend, providing 360° cut protection and a reliable barrier against harmful liquids and chemicals, including sulfuric acid. The gloves feature an anatomical TPR design for impact protection and a dual-coated sandy nitrile palm that ensures superior wet grip performance.
  • Meets ANSI/ISEA A4 cut resistance and Level 4 abrasion resistance standards
  • Impact protection meets ANSI 138 Level 2 and EN 388 impact standards
  • Extended gauntlet cuffs for easy on and off
  • Fully coated in nitrile for protection against oils, solvents, acids, and caustics
  • Machine washable for easy maintenance
  • Includes EN ISO 374-1 and 374-5 protection ratings
Catalog No. 50-006-84868 Supplier MSC Supplier No. 05904180
These chemical-resistant impact gloves are constructed from a high-dexterity 18-gauge HPPE and glass fiber blend, providing 360° cut protection and a reliable barrier against harmful liquids and chemicals, including sulfuric acid. The gloves feature an anatomical TPR design for impact protection and a dual-coated sandy nitrile palm that ensures superior wet grip performance.
  • Meets ANSI/ISEA A4 cut resistance and Level 4 abrasion resistance standards
  • Impact protection meets ANSI 138 Level 2 and EN 388 impact standards
  • Extended gauntlet cuffs for easy on and off
  • Fully coated in nitrile for protection against oils, solvents, acids, and caustics
  • Machine washable for easy maintenance
  • Includes EN ISO 374-1 and 374-5 protection ratings