Learn More
eMolecules​ 89466-08-0 | 2-Hydroxyphenylboronic Acid | Accela ChemBio (ASD) | MFCD01074581 | 137.930 | C6H7BO3 | 95.000 | OB(O)c1ccccc1O | 10g | 447990859
Catalog No. 5000162552 Shop All eMolecules Inc. Products
Encompass_Preferred
2-Hydroxyphenylboronic Acid | Accela ChemBio (ASD) | 89466-08-0 | MFCD01074581 | 137.930 | C6H7BO3 | 95.000 | OB(O)c1ccccc1O | 10g | 447990859
Catalog No. 50-001-62552 Supplier eMolecules​ Supplier No. 447990859
2-Hydroxyphenylboronic Acid | Accela ChemBio (ASD) | 89466-08-0 | MFCD01074581 | 137.930 | C6H7BO3 | 95.000 | OB(O)c1ccccc1O | 10g | 447990859