Learn More
MSC 8979 48X54.8MM BLK DUCT TA 2PK
Catalog No. 501149708
Encompass
This black duct tape is constructed from polyethylene film, fabric, and a rubber adhesive, designed for industrial applications. It offers excellent adhesion to various materials and rough surfaces, suitable for both indoor and outdoor use. With a thickness of 0.29 mm, the tape can be easily unwound, hand-torn, and removed cleanly without residue, even after extended exposure to UV and outdoor weathering.
  • Excellent outdoor UV resistance
  • Removes cleanly for up to six months from most opaque surfaces
  • Sticks immediately and remains for up to one year without deterioration
  • Strong water-resistant backing resists wear and light abrasion
  • Provides a tight, moisture-resistant seal in many containment situations
  • Conforms to irregular surfaces without lifting
  • Meets UL 723 and ASTM E-84 standards
  • Can withstand temperatures up to 93.33 °C (200 °F) for 30 minutes
Catalog No. 50-114-9708 Supplier MSC Supplier No. 53338265
This black duct tape is constructed from polyethylene film, fabric, and a rubber adhesive, designed for industrial applications. It offers excellent adhesion to various materials and rough surfaces, suitable for both indoor and outdoor use. With a thickness of 0.29 mm, the tape can be easily unwound, hand-torn, and removed cleanly without residue, even after extended exposure to UV and outdoor weathering.
  • Excellent outdoor UV resistance
  • Removes cleanly for up to six months from most opaque surfaces
  • Sticks immediately and remains for up to one year without deterioration
  • Strong water-resistant backing resists wear and light abrasion
  • Provides a tight, moisture-resistant seal in many containment situations
  • Conforms to irregular surfaces without lifting
  • Meets UL 723 and ASTM E-84 standards
  • Can withstand temperatures up to 93.33 °C (200 °F) for 30 minutes