Learn More
MSC Duct Tape, 3M, 3 in x 54.8 m, 1 Roll
Catalog No. 501299915
Encompass
This duct tape from 3M Series 8979N features a polyethylene foam material with a rubber adhesive. It has a thickness of 12.1 mil, a width of 3 in., and a length of 54.8 meters, offering a tensile strength of 36 lb/in. This blue tape is suitable for various applications requiring resistance to UV and weather.
  • Resists UV, sunlight, and weather for up to one year without deterioration
  • Available in distinctive red and slate blue colors
  • Complies with MIL-STD-2041D(SH), Notice 2 for leachable halides and sulfur
  • Features a curl-resistant, moisture-resistant, and conformable backing for easy handling
  • Removes cleanly from most opaque surfaces after up to six months of UV exposure and weather
Catalog No. 50-129-9915 Supplier MSC Supplier No. 04583209
This duct tape from 3M Series 8979N features a polyethylene foam material with a rubber adhesive. It has a thickness of 12.1 mil, a width of 3 in., and a length of 54.8 meters, offering a tensile strength of 36 lb/in. This blue tape is suitable for various applications requiring resistance to UV and weather.
  • Resists UV, sunlight, and weather for up to one year without deterioration
  • Available in distinctive red and slate blue colors
  • Complies with MIL-STD-2041D(SH), Notice 2 for leachable halides and sulfur
  • Features a curl-resistant, moisture-resistant, and conformable backing for easy handling
  • Removes cleanly from most opaque surfaces after up to six months of UV exposure and weather