Learn More
MSC 8979N 72X54.8MM RED NUCLEAR DU
Catalog No. 501299917
Encompass
This duct tape features a rubber adhesive and is made of polyethylene foam, designed for high-performance applications. It offers durability and clean removal, making it suitable for various industrial tasks.
  • Resists UV, sunlight, and weather for up to one year without deterioration
  • Complies with MIL-STD-2041D(SH), Notice 2 for leachable halides and sulfur
  • Features a curl-resistant, moisture-resistant, and conformable backing for easy handling
  • Removes cleanly from most opaque surfaces after up to six months of UV exposure
  • Suitable for bundling, tabbing, splicing, pipe bundling, box closing, protecting, holding, reinforcing, joining, identifying, and sealing
  • Tensile strength of 36 lb/in
  • Withstands maximum operating temperatures up to 93.33 °C (200 °F)
  • Meets UL 723, ASME NQA-1, MIL-S-2041D(SH), and ASTM E-84 standards
Catalog No. 50-129-9917 Supplier MSC Supplier No. 53338240
This duct tape features a rubber adhesive and is made of polyethylene foam, designed for high-performance applications. It offers durability and clean removal, making it suitable for various industrial tasks.
  • Resists UV, sunlight, and weather for up to one year without deterioration
  • Complies with MIL-STD-2041D(SH), Notice 2 for leachable halides and sulfur
  • Features a curl-resistant, moisture-resistant, and conformable backing for easy handling
  • Removes cleanly from most opaque surfaces after up to six months of UV exposure
  • Suitable for bundling, tabbing, splicing, pipe bundling, box closing, protecting, holding, reinforcing, joining, identifying, and sealing
  • Tensile strength of 36 lb/in
  • Withstands maximum operating temperatures up to 93.33 °C (200 °F)
  • Meets UL 723, ASME NQA-1, MIL-S-2041D(SH), and ASTM E-84 standards