Learn More
eMolecules​ 898747-24-5 | Methyl 5-bromo-1H-indazole-7-carboxylate | Apollo Scientific US - Building Blocks | MFCD08458991 | 255.071 | C9H7BrN2O2 | 95.000 | COC(=O)c1cc(Br)cc2cn[nH]c12 | 1g | 534604932
Catalog No. 5000164663 Shop All eMolecules Inc. Products
Encompass_Preferred
Methyl 5-bromo-1H-indazole-7-carboxylate | Apollo Scientific US - Building Blocks | 898747-24-5 | MFCD08458991 | 255.071 | C9H7BrN2O2 | 95.000 | COC(=O)c1cc(Br)cc2cn[nH]c12 | 1g | 534604932
Catalog No. 50-001-64663 Supplier eMolecules​ Supplier No. 534604932
Methyl 5-bromo-1H-indazole-7-carboxylate | Apollo Scientific US - Building Blocks | 898747-24-5 | MFCD08458991 | 255.071 | C9H7BrN2O2 | 95.000 | COC(=O)c1cc(Br)cc2cn[nH]c12 | 1g | 534604932