Learn More
Cayman Chemical 8ahydroxy9THCexempt pre 100ug
SDP
Catalog No. 502943275
Encompass_Preferred
An analytical reference standard categorized as a phytocannabinoid; a metabolite of Δ9-THC; has been found in Cannabis; provided as an exempt preparation; intended for research and forensic applications
Catalog No. 50-294-3275 Supplier Cayman Chemical Supplier No. 39793100 UG
An analytical reference standard categorized as a phytocannabinoid; a metabolite of Δ9-THC; has been found in Cannabis; provided as an exempt preparation; intended for research and forensic applications