Learn More
Cayman Chemical 8-Azidoadenosine-5'-O-triphosphate (sodium salt), Purity: ≥95%, 2.7 mg
SDP
Catalog No. 502918784
Encompass_Preferred
A clickable form of ATP; has been used in the synthesis of fluorescent probes for live cell imaging; radiolabeled forms have been used as photoaffinity labels for various proteins, including viral RNA polymerase and Kir6.2
Catalog No. 50-291-8784 Supplier Cayman Chemical Supplier No. 385482.7 MG
A clickable form of ATP; has been used in the synthesis of fluorescent probes for live cell imaging; radiolabeled forms have been used as photoaffinity labels for various proteins, including viral RNA polymerase and Kir6.2