Learn More
Cayman Chemical 8-bromo Cyclic GMP (sodium salt), Purity: ≥98%, CAS: 51116-01-9, 10 mg
SDP
Catalog No. 502968573
Encompass_Preferred
A cell-permeable analog of cGMP that preferentially activates cGMP-dependent protein kinase; 1 µM-0.1 mM inhibits acetylcholine-induced increases in intracellular calcium concentrations
Catalog No. 50-296-8573 Supplier Cayman Chemical Supplier No. 1599210 MG
A cell-permeable analog of cGMP that preferentially activates cGMP-dependent protein kinase; 1 µM-0.1 mM inhibits acetylcholine-induced increases in intracellular calcium concentrations