Learn More
Cayman Chemical 8Bu 3ynTNAD sodIum sal 700ug
SDP
Catalog No. 502916799
Encompass_Preferred
A clickable form of NAD+; has been used in the determination of PARP1-, PARP2-, and PARP3-dependent genomic ADP-ribosylation sites and PARP7 protein substrates using analog-sensitive gatekeeper mutants of these proteins
Catalog No. 50-291-6799 Supplier Cayman Chemical Supplier No. 38477700 UG
A clickable form of NAD+; has been used in the determination of PARP1-, PARP2-, and PARP3-dependent genomic ADP-ribosylation sites and PARP7 protein substrates using analog-sensitive gatekeeper mutants of these proteins