Learn More
USA Scientific Inc 8-cap strip for 0.2 mL tubes
Supplier:  USA Scientific Inc 14001808
Encompass
8-cap strip for 0.2 mL PCR tubes and plates, dome top, mixed colors, 125 strips (1000 caps) per pack. Polypropylene.
Catalog No. 50-274-5434