Learn More
Waters Corp Reservoir Cooled Domino, 8-Channel, Passive Cooling, 4°C
Catalog No. NC3597594 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
The 8-Channel Reservoir Cooled Domino interfaces various consumables with the Andrew+ pipetting robot. It securely holds specific 8-channel reservoirs, providing optimal passive cooling to maintain contents at approximately 4°C for about 5 hours, enhancing diverse laboratory workflows.
  • Provides an interface between consumables and pipetting robot.
  • Securely holds specific 8-channel reservoirs.
  • Utilizes passive cooling technology for optimal cooling efficiency.
  • Maintains reservoir contents at approximately 4°C for about 5 hours.
  • Accommodates up to two 10 mL reservoirs.
  • Equipped with QR codes for critical labware information.
  • Features ARUCO codes for spatial positioning.
  • Enhances automation of diverse laboratory workflows.
Catalog No. NC3597594 Supplier Waters Corp Supplier No. 186010169
The 8-Channel Reservoir Cooled Domino interfaces various consumables with the Andrew+ pipetting robot. It securely holds specific 8-channel reservoirs, providing optimal passive cooling to maintain contents at approximately 4°C for about 5 hours, enhancing diverse laboratory workflows.
  • Provides an interface between consumables and pipetting robot.
  • Securely holds specific 8-channel reservoirs.
  • Utilizes passive cooling technology for optimal cooling efficiency.
  • Maintains reservoir contents at approximately 4°C for about 5 hours.
  • Accommodates up to two 10 mL reservoirs.
  • Equipped with QR codes for critical labware information.
  • Features ARUCO codes for spatial positioning.
  • Enhances automation of diverse laboratory workflows.