Learn More
Cayman Chemical 8CPTCyclIc AMPsodIM salt 10mg
SDP
Catalog No. 502954954
Encompass_Preferred
A lipophilic activator of PKA and PKG (Ka = 0.05 and 0.11 μM, respectively); inhibits PDE VA, PDE III, and PDE IV with IC50 values of 0.9, 24, and 25 μM, respectively
Catalog No. 50-295-4954 Supplier Cayman Chemical Supplier No. 1201110 MG
A lipophilic activator of PKA and PKG (Ka = 0.05 and 0.11 μM, respectively); inhibits PDE VA, PDE III, and PDE IV with IC50 values of 0.9, 24, and 25 μM, respectively