Learn More
Cayman Chemical 8-CPT-Cyclic AMP (sodium salt), Purity: ≥98%, CAS: 93882-12-3, 50 mg
SDP
Catalog No. 502953907
Encompass_Preferred
A lipophilic activator of PKA and PKG (Ka = 0.05 and 0.11 μM, respectively); inhibits PDE VA, PDE III, and PDE IV with IC50 values of 0.9, 24, and 25 μM, respectively
Catalog No. 50-295-3907 Supplier Cayman Chemical Supplier No. 1201150 MG
A lipophilic activator of PKA and PKG (Ka = 0.05 and 0.11 μM, respectively); inhibits PDE VA, PDE III, and PDE IV with IC50 values of 0.9, 24, and 25 μM, respectively