Learn More
Cayman Chemical 8-Cyclopentyl-1,3-dimethylxanthine, Purity: ≥98%, CAS: 35873-49-5, 50 mg
SDP
Catalog No. 502988316
Encompass_Preferred
An adenosine A1 receptor antagonist (Ki = 10.9 nM); selective for adenosine A1 over A2 receptors (Ki = 1,440 nM); inhibits hypoxia-induced depression of evoked synaptic potentials in rat hippocampal slices; increases secondary after-discharge duration in electrically induced seizures in rats ≥2.5 mg/kg; increases the response rate under a fixed-interval schedule of stimulus-shock termination in squirrel monkeys (ED50 = 2.5 μmol/kg)
Catalog No. 50-298-8316 Supplier Cayman Chemical Supplier No. 2987050 MG
An adenosine A1 receptor antagonist (Ki = 10.9 nM); selective for adenosine A1 over A2 receptors (Ki = 1,440 nM); inhibits hypoxia-induced depression of evoked synaptic potentials in rat hippocampal slices; increases secondary after-discharge duration in electrically induced seizures in rats ≥2.5 mg/kg; increases the response rate under a fixed-interval schedule of stimulus-shock termination in squirrel monkeys (ED50 = 2.5 μmol/kg)