Learn More
Cayman Chemical 8dImethylamIno NAD sodIM 700ug
SDP
Catalog No. 502916729
Encompass_Preferred
A derivative of NAD+; has been used in the screening of analog-sensitive PARP1 gatekeeper mutations
Catalog No. 50-291-6729 Supplier Cayman Chemical Supplier No. 38482700 UG
A derivative of NAD+; has been used in the screening of analog-sensitive PARP1 gatekeeper mutations