Learn More
Cayman Chemical 8hydroxy9THCexempt pre 100ug
SDP
Catalog No. 502942073
Encompass_Preferred
An analytical reference standard categorized as a phytocannabinoid metabolite; a metabolite of Δ9-THC; does not induce catalepsy, hypothermia, antinociception in the mouse tetrad assay for cannabimimetic activity and induces hyperlocomotor, rather than hypolocomotor, activity; provided as an exempt preparation; intended for research and forensic applications
Catalog No. 50-294-2073 Supplier Cayman Chemical Supplier No. 39794100 UG
An analytical reference standard categorized as a phytocannabinoid metabolite; a metabolite of Δ9-THC; does not induce catalepsy, hypothermia, antinociception in the mouse tetrad assay for cannabimimetic activity and induces hyperlocomotor, rather than hypolocomotor, activity; provided as an exempt preparation; intended for research and forensic applications