Learn More
Cayman Chemical 8hydroxyexoTHCexempt pr 100ug
SDP
Catalog No. 502943124
Encompass_Preferred
An analytical reference standard that is structurally similar to known phytocannabinoids; a metabolite of exo-THC; provided as an exempt preparation; intended for research and forensic applications
Catalog No. 50-294-3124 Supplier Cayman Chemical Supplier No. 39796100 UG
An analytical reference standard that is structurally similar to known phytocannabinoids; a metabolite of exo-THC; provided as an exempt preparation; intended for research and forensic applications