Learn More
California Pacific Lab & Consulting 8IN ECO FUNL 2IN NPT CRSE THRD
SDP
Catalog No. NC9748958
Encompass_Preferred
8 in. SAFETY ECO FUNNEL FOR LIQUID WASTE COLLECTION, FITS 2 NPT COARSE THRD DRUM BUNG, PREVENTS 99.9% EMISSIONS OF TOXIC FUMES, HDPE, RED LID WITH GASKET THAT LATCHES, REMOVABLE FILTER FOR LARGE PARTICLES, 2000 ML CAPACITY, LIQUID LEVEL TUBE TO PREVENT OV
Catalog No. NC9748958 Supplier California Pacific Lab & Consulting Supplier No. EF47171C
8 in. SAFETY ECO FUNNEL FOR LIQUID WASTE COLLECTION, FITS 2 NPT COARSE THRD DRUM BUNG, PREVENTS 99.9% EMISSIONS OF TOXIC FUMES, HDPE, RED LID WITH GASKET THAT LATCHES, REMOVABLE FILTER FOR LARGE PARTICLES, 2000 ML CAPACITY, LIQUID LEVEL TUBE TO PREVENT OV