Learn More
California Pacific Lab & Consulting 8" ECO Funnel with 100mm cap adapter (Nalgene)
SDP
Catalog No. NC1731844
Encompass_Preferred
This 8" ECO Funnel is designed to fit a 9 liter Nalgene container with a 100mm closure. Our patented design features a latching lid with gasket to keep fumes contained. A removable large particle filter catches stir bars or other items. A detachable tube extends into the waste container to minimize chemical evaporation. The 8" version of our ECO Funnel includes an additional safety feature to prevent over-filling; a secondary air-lock tube creates a small air gap in your waste container, which causes fluid to back up temporarily into the funnel when the container is full. After the screw cap adapter is loosened, the remaining fluid will safely flow into the container and not spill.
Catalog No. NC1731844 Supplier California Pacific Lab & Consulting Supplier No. EF3009
This 8" ECO Funnel is designed to fit a 9 liter Nalgene container with a 100mm closure. Our patented design features a latching lid with gasket to keep fumes contained. A removable large particle filter catches stir bars or other items. A detachable tube extends into the waste container to minimize chemical evaporation. The 8" version of our ECO Funnel includes an additional safety feature to prevent over-filling; a secondary air-lock tube creates a small air gap in your waste container, which causes fluid to back up temporarily into the funnel when the container is full. After the screw cap adapter is loosened, the remaining fluid will safely flow into the container and not spill.