Learn More
Sklar Instruments Forceps, Econo Dressing Forceps, Sklar 8 in., Straight, Serrated, Stainless Steel, BX/12.
SDP
Catalog No. 501098098 Shop All Sklar Instruments Products
Encompass_Preferred
Forceps, Econo Dressing Forceps, Sklar, 8 in., Straight, Serrated, Stainless Steel, BX/12. Econo Dressing Forceps are thumb forceps used for holding dressing materials such as cotton and gauze during surgical procedures, changing dressings, or packing wounds. Econo instruments are Sklar's floor-grade, disposable instruments and are sold in bulk quantities. This product is straight with serrated tips and a length of 8 inches. They are sold in boxes of 12.
Catalog No. 50-109-8098 Supplier Sklar Instruments Supplier No. 21736
Forceps, Econo Dressing Forceps, Sklar, 8 in., Straight, Serrated, Stainless Steel, BX/12. Econo Dressing Forceps are thumb forceps used for holding dressing materials such as cotton and gauze during surgical procedures, changing dressings, or packing wounds. Econo instruments are Sklar's floor-grade, disposable instruments and are sold in bulk quantities. This product is straight with serrated tips and a length of 8 inches. They are sold in boxes of 12.