Learn More
Sklar Instruments Scissors, Supercut Sklar Metzenbaum-Lahey Dissecting Scissors, Sklar 8 in., Blunt/Blunt, Curved, 1 Knifelike Razor Edge and 1 Micro-serrated Edge, Stainless Steel, Reusable, EA.
SDP
Catalog No. 501093437 Shop All Sklar Instruments Products
Encompass_Preferred
Scissors, Supercut Sklarhone Metzenbaum-Lahey Dissecting Scissors, Sklar, 8 in., Blunt/Blunt, Curved, 1 Knifelike Razor Edge and 1 Micro-serrated Edge, Stainless Steel, Reusable, EA. Supercut Sklarhone Metzenbaum-Lahey Dissecting Scissors are used for dissecting and cutting tissue only. These scissors are not recommended for cutting sutures or drains. Metzenbaum-Lahey scissors tend to be shorter than standard Metzenbaum scissors. Supercut Sklarhone scissors combine one knife-like razor edge and one micro-serrated scissor edge. These scissors can be used to cut both delicate and heavy tissue because of their unique design. The micro-serrations of the Supercut Sklarhone scissors minimize tearing of tissue so cuts and edges are smooth and clean. Sklarhone instruments are identified by dual black finger rings. This product is straight, has one razor edge, one micro-serrated edge, blunt tips, and an overall length of 8 inches.
Catalog No. 50-109-3437 Supplier Sklar Instruments Supplier No. 153411
Scissors, Supercut Sklarhone Metzenbaum-Lahey Dissecting Scissors, Sklar, 8 in., Blunt/Blunt, Curved, 1 Knifelike Razor Edge and 1 Micro-serrated Edge, Stainless Steel, Reusable, EA. Supercut Sklarhone Metzenbaum-Lahey Dissecting Scissors are used for dissecting and cutting tissue only. These scissors are not recommended for cutting sutures or drains. Metzenbaum-Lahey scissors tend to be shorter than standard Metzenbaum scissors. Supercut Sklarhone scissors combine one knife-like razor edge and one micro-serrated scissor edge. These scissors can be used to cut both delicate and heavy tissue because of their unique design. The micro-serrations of the Supercut Sklarhone scissors minimize tearing of tissue so cuts and edges are smooth and clean. Sklarhone instruments are identified by dual black finger rings. This product is straight, has one razor edge, one micro-serrated edge, blunt tips, and an overall length of 8 inches.