Learn More
Gilson Company Inc No. 100 Mesh Sieve, All Stainless, Full Height, 8" diameter
Catalog No. NC1488802
Encompass_Preferred
8in (203mm) diameter ASTM E11 Test Sieve has stainless steel woven-wire cloth with No. 100 (150µm) openings and full height stainless steel frame. The overall height is 2-5/8in and the stacked height is 2-1/8in. This sieve is optionally available fitted with #35 (500µm) stainless steel backing cloth for enhanced durability.
Catalog No. NC1488802 Supplier Gilson Company Inc Supplier No. V8SF #100
8in (203mm) diameter ASTM E11 Test Sieve has stainless steel woven-wire cloth with No. 100 (150µm) openings and full height stainless steel frame. The overall height is 2-5/8in and the stacked height is 2-1/8in. This sieve is optionally available fitted with #35 (500µm) stainless steel backing cloth for enhanced durability.