Learn More
Sklar Instruments Scissors, Sims Scissors, Sklar 8 in., Blunt/Blunt, Curved, Smooth, Stainless Steel, EA.
SDP
Catalog No. 501094010 Shop All Sklar Instruments Products
Encompass_Preferred
Scissors, Sims Scissors, Sklar, 8 in., Blunt/Blunt, Curved, Smooth, Stainless Steel, EA. Sklar's Sims Scissors are a finger ring instrument that feature long blades, primarily used for deep tissue dissection. These scissors are available straight or curved with sharp/sharp, blunt/blunt, or sharp/blunt tips. This product is curved with blunt/blunt tips and 8 inches long.
Catalog No. 50-109-4010 Supplier Sklar Instruments Supplier No. 901380
Scissors, Sims Scissors, Sklar, 8 in., Blunt/Blunt, Curved, Smooth, Stainless Steel, EA. Sklar's Sims Scissors are a finger ring instrument that feature long blades, primarily used for deep tissue dissection. These scissors are available straight or curved with sharp/sharp, blunt/blunt, or sharp/blunt tips. This product is curved with blunt/blunt tips and 8 inches long.