Learn More
Sklar Instruments Scissors, Sims Scissors, Sklar 8 in., Sharp/Sharp, Straight, Smooth, Stainless Steel, EA.
SDP
Catalog No. 501094008 Shop All Sklar Instruments Products
Encompass_Preferred
Scissors, Sims Scissors, Sklar, 8 in., Sharp/Sharp, Straight, Smooth, Stainless Steel, EA. Sklar's Sims Scissors are a finger ring instrument that feature long blades, primarily used for deep tissue dissection. These scissors are available straight or curved with sharp/sharp, blunt/blunt, or sharp/blunt tips. This product is straight with sharp/sharp tips and 8 inches long.
Catalog No. 50-109-4008 Supplier Sklar Instruments Supplier No. 901280
Scissors, Sims Scissors, Sklar, 8 in., Sharp/Sharp, Straight, Smooth, Stainless Steel, EA. Sklar's Sims Scissors are a finger ring instrument that feature long blades, primarily used for deep tissue dissection. These scissors are available straight or curved with sharp/sharp, blunt/blunt, or sharp/blunt tips. This product is straight with sharp/sharp tips and 8 inches long.