Learn More
Cayman Chemical 8-Isoprostane ELISA Kit, 96 solid wells
SDP
Supplier:  Cayman Chemical 51635196SOLIDWELLS
Encompass_Preferred
Cayman’s 8-Isoprostane ELISA Kit is a competitive assay that can be used for quantification of 8-isoprostane in plasma, urine, and other sample matrices. The assay has a range from 0.8-500 pg/ml and a sensitivity (80% B/B0) of approximately 3 pg/ml.
Catalog No. NC0929258