Learn More
Cell Biolabs Inc OxiSelect™ 8-iso-Prostaglandin F2a ELISA Kit, 96 assays
Supplier:  Cell Biolabs Inc STA337
Encompass_Preferred
8-iso-Prostaglandin F2α (also known as 8-epi-PGF2α, 8-isoprostane, or 15-isoprostane F2t), is an isoprostane that has been shown to be useful for the assessment of oxidative stress in vivo. It is produced in membrane phospholipids from non-cyclooxygenase and cyclooxygenase peroxidation pathways derived from arachidonic acid. 8-iso-Prostaglandin F2α (8-iso-PGF2α) is a potent vasoconstrictor, a mutagen in 3T3 cells as well as vascular smooth muscle cells, and also a possible pathophysiological mediator that can alter membrane integrity. The OxiSelect™ 8-iso-Prostaglandin F2α ELISA Kit is an enzyme immunoassay developed for rapid detection and quantification of 8-iso-PGF2α. The quantity of 8-iso-PGF2α in samples is determined by comparing its absorbance with that of a known 8-iso-PGF2α standard curve. Each kit provides sufficient reagents to perform up to 96 assays, including the standard curve and unknown samples.
Catalog No. NC0962418